Product Details

SNP ID
rs138770477
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:50360770 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTTCTTTATAGGGCAGGTGACTT[C/T]CTGCATGGAGGATGCAACTTCTTGT
Phenotype
MIM: 605631
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NAPSA PubMed Links

Gene Details

Gene
NAPSA
Gene Name
napsin A aspartic peptidase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004851.2 Intron NP_004842.1
XM_011527540.1 Intron XP_011525842.1
XM_017027512.1 Intron XP_016883001.1

View Full Product Details