Product Details

SNP ID
rs138845372
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:58469198 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTTAGGACCAGATGGCCTTTGAG[A/G]ATGTGGCTGTGTACTTCTCCCAGGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF324 PubMed Links

Gene Details

Gene
ZNF324
Gene Name
zinc finger protein 324
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014347.2 953 Missense Mutation AAT,GAT N5D NP_055162.1
XM_005258713.4 953 Missense Mutation AAT,GAT N10D XP_005258770.1
Gene
ZNF446
Gene Name
zinc finger protein 446
There are no transcripts associated with this gene.

View Full Product Details