Product Details

SNP ID
rs139213045
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:10833012 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCAGGGGCTACGTGGGCACCGGGC[A/G]CTTGTCCTGGAAGAAGCGCTTGAGC
Phenotype
MIM: 602378 MIM: 605395
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C19orf38 PubMed Links

Gene Details

Gene
C19orf38
Gene Name
chromosome 19 open reading frame 38
There are no transcripts associated with this gene.

Gene
DNM2
Gene Name
dynamin 2
There are no transcripts associated with this gene.

Gene
MIR6793
Gene Name
microRNA 6793
There are no transcripts associated with this gene.

Gene
TMED1
Gene Name
transmembrane p24 trafficking protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006858.3 783 Missense Mutation CGC,TGC R223C NP_006849.1
XM_006722631.3 783 Missense Mutation CGC,TGC R185C XP_006722694.1

View Full Product Details