Product Details

SNP ID
rs139225148
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:2039478 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAAAAACCTTCTATAAAACACCCC[A/C]CTCAGCTGAGCTTAGTGCCTGGGAA
Phenotype
MIM: 605069
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MKNK2 PubMed Links

Gene Details

Gene
MKNK2
Gene Name
MAP kinase interacting serine/threonine kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017572.3 1778 Intron NP_060042.2
NM_199054.2 1778 UTR 3 NP_951009.1

View Full Product Details