Product Details

SNP ID
rs139234673
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:36610662 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCCATCATCAGGGTTTCACATGG[C/T]TAAGCCTGATATGATCCGCAAGTTG
Phenotype
MIM: 609516
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF382 PubMed Links

Gene Details

Gene
ZNF382
Gene Name
zinc finger protein 382
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256838.1 254 Missense Mutation GCT,GTT A50V NP_001243767.1
NM_032825.4 254 Missense Mutation GCT,GTT A51V NP_116214.2
Gene
ZNF529
Gene Name
zinc finger protein 529
There are no transcripts associated with this gene.

View Full Product Details