Product Details

SNP ID
rs139315951
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:32352933 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCAGTCCTTCATTGGAAATTGAT[A/G]AACAAAGAAAAACTAAACCAGATCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF507 PubMed Links

Gene Details

Gene
ZNF507
Gene Name
zinc finger protein 507
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136156.1 281 Missense Mutation AAA,GAA K35E NP_001129628.1
NM_014910.4 281 Missense Mutation AAA,GAA K35E NP_055725.2

View Full Product Details