Product Details
- SNP ID
-
rs139322292
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:56223750 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGTTGGCTGGACACGTCTTTCAGAT[A/C]ATCTCTGACACTGGAGGGGGCTTCA
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ZSCAN5A
PubMed Links
Gene Details
- Gene
- ZSCAN5A
- Gene Name
- zinc finger and SCAN domain containing 5A
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001322061.1 |
818 |
Missense Mutation |
GAT,TAT |
D210Y |
NP_001308990.1 |
NM_001322062.1 |
818 |
Intron |
|
|
NP_001308991.1 |
NM_001322064.1 |
818 |
Missense Mutation |
GAT,TAT |
D157Y |
NP_001308993.1 |
NM_001322065.1 |
818 |
Missense Mutation |
GAT,TAT |
D157Y |
NP_001308994.1 |
NM_001322066.1 |
818 |
Missense Mutation |
GAT,TAT |
D157Y |
NP_001308995.1 |
NM_001322067.1 |
818 |
Missense Mutation |
GAT,TAT |
D157Y |
NP_001308996.1 |
NM_001322068.1 |
818 |
Missense Mutation |
GAT,TAT |
D157Y |
NP_001308997.1 |
NM_001322069.1 |
818 |
Missense Mutation |
GAT,TAT |
D157Y |
NP_001308998.1 |
NM_001322070.1 |
818 |
Missense Mutation |
GAT,TAT |
D157Y |
NP_001308999.1 |
NM_001322072.1 |
818 |
Missense Mutation |
GAT,TAT |
D157Y |
NP_001309001.1 |
NM_001322073.1 |
818 |
Missense Mutation |
GAT,TAT |
D157Y |
NP_001309002.1 |
NM_001322074.1 |
818 |
Missense Mutation |
GAT,TAT |
D157Y |
NP_001309003.1 |
NM_001322075.1 |
818 |
Missense Mutation |
GAT,TAT |
D157Y |
NP_001309004.1 |
NM_001322076.1 |
818 |
Missense Mutation |
GAT,TAT |
D157Y |
NP_001309005.1 |
NM_001322077.1 |
818 |
Missense Mutation |
GAT,TAT |
D40Y |
NP_001309006.1 |
NM_001322078.1 |
818 |
Missense Mutation |
GAT,TAT |
D27Y |
NP_001309007.1 |
NM_024303.2 |
818 |
Intron |
|
|
NP_077279.1 |
XM_005259254.1 |
818 |
Intron |
|
|
XP_005259311.1 |
XM_006723377.3 |
818 |
Missense Mutation |
GAT,TAT |
D233Y |
XP_006723440.1 |
XM_011527308.1 |
818 |
Intron |
|
|
XP_011525610.1 |
XM_017027298.1 |
818 |
Missense Mutation |
GAT,TAT |
D233Y |
XP_016882787.1 |
XM_017027299.1 |
818 |
Missense Mutation |
GAT,TAT |
D157Y |
XP_016882788.1 |
View Full Product Details