Product Details

SNP ID
rs139356853
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:814530 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATCTGCGTGAGCCCGCATACGCC[A/C]GCGGCGATGGCAAAGGCGAAGACCA
Phenotype
MIM: 610391 MIM: 600693
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MIR3187 PubMed Links

Gene Details

Gene
MIR3187
Gene Name
microRNA 3187
There are no transcripts associated with this gene.

Gene
MIR4745
Gene Name
microRNA 4745
There are no transcripts associated with this gene.

Gene
PLPPR3
Gene Name
phospholipid phosphatase related 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270366.1 1475 Silent Mutation NP_001257295.1
NM_024888.2 1475 Silent Mutation NP_079164.1
XM_011528317.2 1475 Silent Mutation XP_011526619.1
Gene
PTBP1
Gene Name
polypyrimidine tract binding protein 1
There are no transcripts associated with this gene.

View Full Product Details