Product Details

SNP ID
rs139419159
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:43718934 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCAGGCTGCCCGGCTGACAAGTAC[C/G]TAAAGCAGGTAGACTTCAGCCGCTA
Phenotype
MIM: 613176
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
IRGC PubMed Links

Gene Details

Gene
IRGC
Gene Name
immunity related GTPase cinema
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019612.3 557 Missense Mutation CTA,GTA L126V NP_062558.1
XM_005259058.3 557 Missense Mutation CTA,GTA L163V XP_005259115.1
XM_006723283.3 557 Missense Mutation CTA,GTA L126V XP_006723346.1
XM_011527118.2 557 Missense Mutation CTA,GTA L126V XP_011525420.1
Gene
SMG9
Gene Name
SMG9, nonsense mediated mRNA decay factor
There are no transcripts associated with this gene.

View Full Product Details