Product Details

SNP ID
rs140375744
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:18281074 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCCAGGGCCTTGACGAAGCCCTC[A/G]GCGAACTCCTGCTCCTCGCTGGCCG
Phenotype
MIM: 165162
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
JUND PubMed Links

Gene Details

Gene
JUND
Gene Name
JunD proto-oncogene, AP-1 transcription factor subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286968.1 583 Silent Mutation NP_001273897.1
NM_005354.5 583 Silent Mutation NP_005345.3
Gene
KIAA1683
Gene Name
KIAA1683
There are no transcripts associated with this gene.

Gene
MIR3188
Gene Name
microRNA 3188
There are no transcripts associated with this gene.

View Full Product Details