Product Details

SNP ID
rs140655170
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:43543662 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGCTGTGATCACAAACTGAACCC[A/G]GTCACTCATATAGTCCTCGAGCTCC
Phenotype
MIM: 194360
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
XRCC1 PubMed Links

Gene Details

Gene
XRCC1
Gene Name
X-ray repair cross complementing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006297.2 1858 Missense Mutation CGG,TGG R580W NP_006288.2
Gene
ZNF575
Gene Name
zinc finger protein 575
There are no transcripts associated with this gene.

View Full Product Details