Product Details

SNP ID
rs113077195
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:54852135 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAAGAAGACAGAAGCCACGTTCCC[A/T]TCTTCCACGGCAGAATATTCCAGGA
Phenotype
MIM: 604955 MIM: 604947
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
KIR2DS4 PubMed Links

Gene Details

Gene
KIR2DS4
Gene Name
killer cell immunoglobulin like receptor, two Ig domains and short cytoplasmic tail 4
There are no transcripts associated with this gene.

Gene
KIR3DL2
Gene Name
killer cell immunoglobulin like receptor, three Ig domains and long cytoplasmic tail 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242867.1 365 Intron NP_001229796.1
NM_006737.3 365 Intron NP_006728.2
XM_011526940.2 365 Missense Mutation ATC,TTC I70F XP_011525242.1
XM_017026784.1 365 Missense Mutation ATC,TTC I70F XP_016882273.1

View Full Product Details