Product Details
- SNP ID
-
rs141066224
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:1397495 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGCCTCCAGCAGCGCGGGCACCTGC[A/G]TCTCCTGGTCGGGGATGGCACCAGG
- Phenotype
-
MIM: 601240
MIM: 601825
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
GAMT
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs17851582] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GAMT
- Gene Name
- guanidinoacetate N-methyltransferase
- Gene
- NDUFS7
- Gene Name
- NADH:ubiquinone oxidoreductase core subunit S7
There are no transcripts associated with this gene.
View Full Product Details