Product Details

SNP ID
rs141315927
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:55624484 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCAGGCCCCTTCCTTGCCCGCAC[C/T]AGGTCCAGTGGCTCCTGGGATCGCG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF784 PubMed Links

Gene Details

Gene
ZNF784
Gene Name
zinc finger protein 784
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_203374.1 92 Silent Mutation CTA,CTG L26L NP_976308.1
Gene
ZNF865
Gene Name
zinc finger protein 865
There are no transcripts associated with this gene.

View Full Product Details