Product Details

SNP ID
rs141501557
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:54252922 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGATGTCGGAGGAGGAGGAAGAGGA[A/G]GAGGAACAGCAGCAGGACGAAGGCC
Phenotype
MIM: 604814
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LILRB5 PubMed Links

Gene Details

Gene
LILRB5
Gene Name
leukocyte immunoglobulin like receptor B5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001081442.2 1511 Missense Mutation CTC,TTC L475F NP_001074911.1
NM_001081443.2 1511 Missense Mutation CTC,TTC L375F NP_001074912.1
NM_001304457.1 1511 Missense Mutation CTC,TTC L466F NP_001291386.1
NM_006840.4 1511 Missense Mutation CTC,TTC L474F NP_006831.1
XM_011526359.2 1511 Missense Mutation CTC,TTC L475F XP_011524661.1
XM_011526360.2 1511 Missense Mutation CTC,TTC L474F XP_011524662.1
XM_011526361.2 1511 Missense Mutation CTC,TTC L458F XP_011524663.1
XM_011526362.2 1511 Missense Mutation CTC,TTC L375F XP_011524664.1

View Full Product Details