Product Details
- SNP ID
-
rs141734238
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:55174970 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCCCGGCCGTGGGGACATAGCGGAG[C/G]GAGAAGCAGATGTCCCCAAGCTTCT
- Phenotype
-
MIM: 614566
MIM: 602510
MIM: 600782
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
DNAAF3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs12459917] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DNAAF3
- Gene Name
- dynein axonemal assembly factor 3
There are no transcripts associated with this gene.
- Gene
- PTPRH
- Gene Name
- protein tyrosine phosphatase, receptor type H
There are no transcripts associated with this gene.
- Gene
- SYT5
- Gene Name
- synaptotagmin 5
View Full Product Details