Product Details
- SNP ID
-
rs141774128
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:51347012 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGGCCCCCATCGATCTCCCGCTCC[A/G]CTTTCAACTTGTCCCCCTCCAGCGT
- Phenotype
-
MIM: 130410
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ETFB
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs144640661] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ETFB
- Gene Name
- electron transfer flavoprotein beta subunit
- Gene
- VSIG10L
- Gene Name
- V-set and immunoglobulin domain containing 10 like
There are no transcripts associated with this gene.
View Full Product Details