Product Details

SNP ID
rs141810971
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:39132819 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCTCACTCCCTTGCTGGAATAGA[C/T]GGGTGTCACCTGACATTCTCACCTT
Phenotype
MIM: 605451
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PAK4 PubMed Links

Gene Details

Gene
PAK4
Gene Name
p21 (RAC1) activated kinase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001014831.2 Intron NP_001014831.1
NM_001014832.1 Intron NP_001014832.1
NM_001014834.2 Intron NP_001014834.1
NM_001014835.1 Intron NP_001014835.1
NM_005884.3 Intron NP_005875.1
XM_011526316.1 Intron XP_011524618.1
XM_011526317.2 Intron XP_011524619.1
XM_011526318.2 Intron XP_011524620.1
XM_011526319.2 Intron XP_011524621.1
XM_011526320.1 Intron XP_011524622.1

View Full Product Details