Product Details

SNP ID
rs141937977
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57193886 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTGACTTTCCAGGTGTCTGTGAC[C/G]TTTGATGATGTGGCTGTGACTTTCA
Phenotype
MIM: 604668
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZNF264 PubMed Links

Gene Details

Gene
ZNF264
Gene Name
zinc finger protein 264
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003417.4 459 Silent Mutation ACC,ACG T15T NP_003408.1
XM_011527522.2 459 Silent Mutation ACC,ACG T15T XP_011525824.1

View Full Product Details