Product Details

SNP ID
rs142492326
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:50906943 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCCAGTCCCCAGAGTTAACTGGC[C/T]TGGACGGTTTTCTCTATCCACTCAG
Phenotype
MIM: 603767
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KLK4 PubMed Links

Gene Details

Gene
KLK4
Gene Name
kallikrein related peptidase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001302961.1 756 Silent Mutation CAA,CAG Q157Q NP_001289890.1
NM_004917.4 756 Silent Mutation CAA,CAG Q252Q NP_004908.4
XM_005259441.4 756 Silent Mutation CAA,CAG Q157Q XP_005259498.2
XM_011527545.2 756 Intron XP_011525847.1
XM_011527546.2 756 UTR 3 XP_011525848.1

View Full Product Details