Product Details
- SNP ID
-
rs142543824
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:9610969 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTGTGAAGAAGCTGTGACAGCTCTC[C/T]CATATTCCTGAAATTCACAGAGTTT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF561
PubMed Links
Gene Details
- Gene
- ZNF561
- Gene Name
- zinc finger protein 561
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_152289.2 |
852 |
Missense Mutation |
GAG,GGG |
E231G |
NP_689502.2 |
XM_005260150.2 |
852 |
Missense Mutation |
GAG,GGG |
E231G |
XP_005260207.1 |
XM_005260151.2 |
852 |
Missense Mutation |
GAG,GGG |
E124G |
XP_005260208.1 |
XM_006722938.2 |
852 |
Missense Mutation |
GAG,GGG |
E237G |
XP_006723001.1 |
XM_011528433.2 |
852 |
Missense Mutation |
GAG,GGG |
E237G |
XP_011526735.1 |
XM_011528434.2 |
852 |
Missense Mutation |
GAG,GGG |
E237G |
XP_011526736.1 |
XM_011528436.2 |
852 |
Missense Mutation |
GAG,GGG |
E204G |
XP_011526738.1 |
XM_011528437.2 |
852 |
Missense Mutation |
GAG,GGG |
E204G |
XP_011526739.1 |
XM_017027478.1 |
852 |
Missense Mutation |
GAG,GGG |
E204G |
XP_016882967.1 |
XM_017027479.1 |
852 |
Missense Mutation |
GAG,GGG |
E162G |
XP_016882968.1 |
XM_017027480.1 |
852 |
Missense Mutation |
GAG,GGG |
E162G |
XP_016882969.1 |
XM_017027481.1 |
852 |
Missense Mutation |
GAG,GGG |
E162G |
XP_016882970.1 |
XM_017027482.1 |
852 |
Missense Mutation |
GAG,GGG |
E95G |
XP_016882971.1 |
View Full Product Details