Product Details

SNP ID
rs142808498
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:6739767 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGGCGGCGGCGGGAGCAGCATGGA[C/T]TGGGGCACTGAGCTGTGGGTAAGTC
Phenotype
MIM: 604504
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GPR108 PubMed Links

Gene Details

Gene
GPR108
Gene Name
G protein-coupled receptor 108
There are no transcripts associated with this gene.

Gene
MIR6791
Gene Name
microRNA 6791
There are no transcripts associated with this gene.

Gene
TRIP10
Gene Name
thyroid hormone receptor interactor 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001288962.1 86 Silent Mutation GAC,GAT D2D NP_001275891.1
NM_001288963.1 86 Silent Mutation GAC,GAT D2D NP_001275892.1
NM_004240.3 86 Silent Mutation GAC,GAT D2D NP_004231.1
XM_005259683.2 86 Silent Mutation GAC,GAT D2D XP_005259740.1
XM_006722940.1 86 Silent Mutation GAC,GAT D2D XP_006723003.1

View Full Product Details