Product Details

SNP ID
rs142842600
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:56568497 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTTCTATGGCTTAATCACGTTCT[A/G]GTCAATAACGGAAGATATCCCCATT
Phenotype
MIM: 616798
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZFP28 PubMed Links

Gene Details

Gene
ZFP28
Gene Name
ZFP28 zinc finger protein
There are no transcripts associated with this gene.

Gene
ZNF470
Gene Name
zinc finger protein 470
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001668.3 Intron NP_001001668.3
XM_017026801.1 Intron XP_016882290.1

View Full Product Details