Product Details
- SNP ID
-
rs143109041
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:39734345 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AATTCTTGGCCATCCTGAAAGGGCA[G/T]ATTCTTGGATTCCACCTGCTGCTTC
- Phenotype
-
MIM: 153310
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CLC
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs384138] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CLC
- Gene Name
- Charcot-Leyden crystal galectin
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001828.5 |
|
Intron |
|
|
NP_001819.2 |
View Full Product Details