Product Details

SNP ID
rs143131074
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:41837145 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTGTCGGGTGCAGCTAGATGGTC[A/G]GTCCCTGAAGGCAAACAGGAGGCCT
Phenotype
MIM: 614806
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DMRTC2 PubMed Links

Gene Details

Gene
DMRTC2
Gene Name
DMRT like family C2
There are no transcripts associated with this gene.

Gene
LYPD4
Gene Name
LY6/PLAUR domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005278385.4 Intron XP_005278442.1
XM_005278387.4 Intron XP_005278444.1

View Full Product Details