Product Details

SNP ID
rs143455686
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:9186180 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTCTGCCACCCTCTGCACTATAT[A/G]ATCATCATGAACCCCCACCTCTGTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR7D2 PubMed Links

Gene Details

Gene
OR7D2
Gene Name
olfactory receptor family 7 subfamily D member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175883.2 587 Missense Mutation ATA,ATG I133M NP_787079.1

View Full Product Details