Product Details

SNP ID
rs143462258
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:7070563 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATTTTTGTGCCCTTTTTATTTAGA[C/T]GAAAAAAGTATAATTCAAGCTCAGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF557 PubMed Links

Gene Details

Gene
ZNF557
Gene Name
zinc finger protein 557
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001044387.1 316 UTR 5 NP_001037852.1
NM_001044388.1 316 UTR 5 NP_001037853.1
NM_024341.2 316 UTR 5 NP_077317.2

View Full Product Details