Product Details

SNP ID
rs143606600
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:44916271 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTTTGGAAACACACTGGAGGACA[A/G]GGCTCGGGAACTCATCAGCCGCATC
Phenotype
MIM: 107710 MIM: 107741
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
APOC1 PubMed Links

Gene Details

Gene
APOC1
Gene Name
apolipoprotein C1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321065.1 339 Missense Mutation AAG,AGG K47R NP_001307994.1
NM_001321066.1 339 Missense Mutation AAG,AGG K47R NP_001307995.1
NM_001645.4 339 Missense Mutation AAG,AGG K47R NP_001636.1
Gene
APOE
Gene Name
apolipoprotein E
There are no transcripts associated with this gene.

View Full Product Details