Product Details

SNP ID
rs143694151
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:38946485 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCTCACTCACCAGTCAGCCACAC[A/G]GATCTCAATCTGGGCGCTGTCCAGC
Phenotype
MIM: 609094
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FBXO17 PubMed Links

Gene Details

Gene
FBXO17
Gene Name
F-box protein 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024907.6 676 Missense Mutation CGT,TGT R182C NP_079183.4
NM_148169.2 676 Missense Mutation CGT,TGT R191C NP_680474.1

View Full Product Details