Product Details

SNP ID
rs143888878
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:45072118 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCAGCAGCATGGACAGCCGGCTCC[A/G]GAGGGGCTGCAGAGGCCTGCTCCTG
Phenotype
MIM: 607419 MIM: 613226
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLASRP PubMed Links

Gene Details

Gene
CLASRP
Gene Name
CLK4 associating serine/arginine rich protein
There are no transcripts associated with this gene.

Gene
GEMIN7
Gene Name
gem nuclear organelle associated protein 7
There are no transcripts associated with this gene.

Gene
ZNF296
Gene Name
zinc finger protein 296
There are no transcripts associated with this gene.

View Full Product Details