Product Details

SNP ID
rs144106235
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:39408124 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATGGCCTGGGCGAGCTGCGCCAG[A/G]CCAATCGCCACCCCAAATACAAGAC
Phenotype
MIM: 612914 MIM: 611893 MIM: 190700
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MED29 PubMed Links

Gene Details

Gene
MED29
Gene Name
mediator complex subunit 29
There are no transcripts associated with this gene.

Gene
MIR4530
Gene Name
microRNA 4530
There are no transcripts associated with this gene.

Gene
PLEKHG2
Gene Name
pleckstrin homology and RhoGEF domain containing G2
There are no transcripts associated with this gene.

Gene
ZFP36
Gene Name
ZFP36 ring finger protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003407.3 464 Missense Mutation ACC,GCC T142A NP_003398.2

View Full Product Details