Product Details

SNP ID
rs144130932
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:828263 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCTTTTGGACATCGTTGGCGGCC[A/G]GAAGGCGAGGCCCCGCCAGTTCCCG
Phenotype
MIM: 162815 MIM: 610391
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AZU1 PubMed Links

Gene Details

Gene
AZU1
Gene Name
azurocidin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001700.4 113 Missense Mutation CAG,CGG Q31R NP_001691.1
Gene
PLPPR3
Gene Name
phospholipid phosphatase related 3
There are no transcripts associated with this gene.

View Full Product Details