Product Details

SNP ID
rs144230339
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:6743077 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGTCCGTGTATGTCTTGAGCTGA[C/T]CAAGTACTCACAAGAGATGAAACAG
Phenotype
MIM: 604721 MIM: 604504
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GPR108 PubMed Links

Gene Details

Gene
GPR108
Gene Name
G protein-coupled receptor 108
There are no transcripts associated with this gene.

Gene
MIR6791
Gene Name
microRNA 6791
There are no transcripts associated with this gene.

Gene
SH2D3A
Gene Name
SH2 domain containing 3A
There are no transcripts associated with this gene.

Gene
TRIP10
Gene Name
thyroid hormone receptor interactor 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001288962.1 388 Missense Mutation ACC,ATC T103I NP_001275891.1
NM_001288963.1 388 Missense Mutation ACC,ATC T103I NP_001275892.1
NM_004240.3 388 Missense Mutation ACC,ATC T103I NP_004231.1
XM_005259683.2 388 Missense Mutation ACC,ATC T103I XP_005259740.1
XM_006722940.1 388 Missense Mutation ACC,ATC T103I XP_006723003.1

View Full Product Details