Product Details

SNP ID
rs144390977
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:6306804 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCAGTAGCGGACTTTGAGGGTTTC[A/C]CCTGGCATCTCATAGTTGGCATCCA
Phenotype
MIM: 613491
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
ACER1 PubMed Links

Gene Details

Gene
ACER1
Gene Name
alkaline ceramidase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_133492.2 783 Silent Mutation GGG,GGT G235G NP_597999.1
XM_011527673.2 783 Silent Mutation GGG,GGT G192G XP_011525975.1

View Full Product Details