Product Details

SNP ID
rs144698189
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:9115105 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTCTCAGAACAGAGGTGACTATT[G/T]GAGAATAAGAGAAAATTATTCCAGA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
OR7G1 PubMed Links

Gene Details

Gene
OR7G1
Gene Name
olfactory receptor family 7 subfamily G member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005192.2 659 Missense Mutation CAA,CCA Q220P NP_001005192.2

View Full Product Details