Product Details

SNP ID
rs145079363
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:50908697 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGACACGGATTCGTCCAACTTGAT[A/G]AGCATGAGGTCGTTAGCGAGCAAGG
Phenotype
MIM: 603767
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KLK4 PubMed Links

Gene Details

Gene
KLK4
Gene Name
kallikrein related peptidase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001302961.1 357 Silent Mutation CTC,CTT L24L NP_001289890.1
NM_004917.4 357 Silent Mutation CTC,CTT L119L NP_004908.4
XM_005259441.4 357 Silent Mutation CTC,CTT L24L XP_005259498.2
XM_011527545.2 357 Silent Mutation CTC,CTT L119L XP_011525847.1
XM_011527546.2 357 Silent Mutation CTC,CTT L119L XP_011525848.1

View Full Product Details