Product Details

SNP ID
rs145299279
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:846254 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGCCTGGCCATGGGCTGGGGCCG[C/T]GTGGGTGCCCACGACCCCCCAGCCC
Phenotype
MIM: 130130 MIM: 177020
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ELANE PubMed Links

Gene Details

Gene
ELANE
Gene Name
elastase, neutrophil expressed
There are no transcripts associated with this gene.

Gene
PRTN3
Gene Name
proteinase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002777.3 521 Silent Mutation CGC,CGT R159R NP_002768.3
XM_011528136.1 521 Silent Mutation CGC,CGT R159R XP_011526438.1

View Full Product Details