Product Details
- SNP ID
-
rs145428670
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:5810607 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATAGCCAACATTGCTGGTGTTGTTA[C/T]TGTTGTTAGGCCAGAAAGCACGTGG
- Phenotype
-
MIM: 602018
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NRTN
PubMed Links
Gene Details
- Gene
- NRTN
- Gene Name
- neurturin
View Full Product Details