Product Details

SNP ID
rs145507498
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:4548365 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACGAGGAACTTGAGGACCGTCCCC[A/G]CCTCAGAACCCAGGAAGACAACGGT
Phenotype
MIM: 611289 MIM: 608873
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LRG1 PubMed Links

Gene Details

Gene
LRG1
Gene Name
leucine rich alpha-2-glycoprotein 1
There are no transcripts associated with this gene.

Gene
SEMA6B
Gene Name
semaphorin 6B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032108.3 1614 Missense Mutation GCG,GTG A451V NP_115484.2
XM_011527639.2 1614 Missense Mutation GCG,GTG A451V XP_011525941.1
XM_011527640.2 1614 Missense Mutation GCG,GTG A451V XP_011525942.1
XM_011527641.1 1614 Missense Mutation GCG,GTG A451V XP_011525943.1

View Full Product Details