Product Details

SNP ID
rs145674301
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:6309753 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGCAGGGCAATGCTGTTGAGGGC[A/G]TAGGCGTTGACCGTGGGCCGCAGGA
Phenotype
MIM: 613491
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACER1 PubMed Links

Gene Details

Gene
ACER1
Gene Name
alkaline ceramidase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_133492.2 510 Silent Mutation NP_597999.1
XM_011527673.2 510 Silent Mutation XP_011525975.1

View Full Product Details