Product Details

SNP ID
rs145704690
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:43597145 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAACCCTGAAGAGAACATGAAGCAG[C/T]AGGATTCACCCAAGGAGAGAAGTCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IRGQ PubMed Links

Gene Details

Gene
IRGQ
Gene Name
immunity related GTPase Q
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007561.2 210 Intron NP_001007562.1
XM_005258515.3 210 Intron XP_005258572.1
Gene
ZNF576
Gene Name
zinc finger protein 576
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145347.1 210 Nonsense Mutation CAG,TAG Q13* NP_001138819.1
NM_024327.2 210 Nonsense Mutation CAG,TAG Q13* NP_077303.1

View Full Product Details