Product Details

SNP ID
rs145967244
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:2243532 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCGGCAGCTGGCCCTGGAGACCAT[C/T]GACATCAACAAGGTGGGCCAGCCAC
Phenotype
MIM: 600957 MIM: 608743 MIM: 600796
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AMH PubMed Links

Gene Details

Gene
AMH
Gene Name
anti-Mullerian hormone
There are no transcripts associated with this gene.

Gene
JSRP1
Gene Name
junctional sarcoplasmic reticulum protein 1
There are no transcripts associated with this gene.

Gene
MIR1227
Gene Name
microRNA 1227
There are no transcripts associated with this gene.

Gene
MIR4321
Gene Name
microRNA 4321
There are no transcripts associated with this gene.

Gene
MIR6789
Gene Name
microRNA 6789
There are no transcripts associated with this gene.

Gene
PLEKHJ1
Gene Name
pleckstrin homology domain containing J1
There are no transcripts associated with this gene.

Gene
SF3A2
Gene Name
splicing factor 3a subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007165.4 236 Silent Mutation ATC,ATT I38I NP_009096.2

View Full Product Details