Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001136139.2 | 2052 | Missense Mutation | NP_001129611.1 | ||
NM_003200.3 | 2052 | Missense Mutation | NP_003191.1 | ||
XM_006722852.3 | 2052 | Missense Mutation | XP_006722915.1 | ||
XM_006722853.3 | 2052 | Missense Mutation | XP_006722916.1 | ||
XM_006722855.3 | 2052 | Missense Mutation | XP_006722918.1 | ||
XM_006722856.3 | 2052 | Missense Mutation | XP_006722919.1 | ||
XM_006722857.3 | 2052 | Missense Mutation | XP_006722920.1 | ||
XM_006722858.3 | 2052 | Missense Mutation | XP_006722921.1 | ||
XM_011528215.2 | 2052 | Missense Mutation | XP_011526517.1 | ||
XM_011528216.2 | 2052 | Missense Mutation | XP_011526518.1 | ||
XM_011528217.2 | 2052 | Missense Mutation | XP_011526519.1 | ||
XM_011528219.2 | 2052 | Missense Mutation | XP_011526521.1 | ||
XM_011528220.2 | 2052 | Missense Mutation | XP_011526522.1 | ||
XM_011528221.2 | 2052 | Missense Mutation | XP_011526523.1 | ||
XM_011528223.2 | 2052 | Missense Mutation | XP_011526525.1 | ||
XM_011528225.2 | 2052 | Missense Mutation | XP_011526527.1 | ||
XM_011528226.2 | 2052 | Missense Mutation | XP_011526528.1 | ||
XM_011528227.2 | 2052 | Missense Mutation | XP_011526529.1 | ||
XM_017027177.1 | 2052 | Missense Mutation | XP_016882666.1 | ||
XM_017027178.1 | 2052 | Missense Mutation | XP_016882667.1 | ||
XM_017027179.1 | 2052 | Missense Mutation | XP_016882668.1 | ||
XM_017027180.1 | 2052 | Missense Mutation | XP_016882669.1 | ||
XM_017027181.1 | 2052 | Missense Mutation | XP_016882670.1 | ||
XM_017027182.1 | 2052 | Missense Mutation | XP_016882671.1 | ||
XM_017027183.1 | 2052 | Missense Mutation | XP_016882672.1 | ||
XM_017027184.1 | 2052 | Intron | XP_016882673.1 |