Product Details

SNP ID
rs146176366
Assay Type
Functionally Tested
NCBI dbSNP Submissions
4
Location
Chr.19:49365644 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCCTCTCCAGCCACCTCCAGATC[A/G]ACAAGGTGCCTATGCAGGGAAGCCC
Phenotype
MIM: 605418 MIM: 601729
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
DKKL1 PubMed Links

Gene Details

Gene
DKKL1
Gene Name
dickkopf like acrosomal protein 1
There are no transcripts associated with this gene.

Gene
LOC101928295
Gene Name
uncharacterized LOC101928295
There are no transcripts associated with this gene.

Gene
TEAD2
Gene Name
TEA domain transcription factor 2
There are no transcripts associated with this gene.

View Full Product Details