Product Details
- SNP ID
-
rs146361951
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:58238478 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCAGGCTTGTGGAGCGGTGCATAGT[A/G]CCTGGAAGACGGGGACACAGGGTTT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF544
PubMed Links
Gene Details
- Gene
- ZNF544
- Gene Name
- zinc finger protein 544
View Full Product Details