Product Details

SNP ID
rs146517398
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:12623723 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGACTCAGTGGCTTTTGAGGATGT[A/G]TCTGTGAGCTTCAGCCAGGAGGAGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF791 PubMed Links

Gene Details

Gene
ZNF791
Gene Name
zinc finger protein 791
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153358.2 159 Silent Mutation GTA,GTG V9V NP_699189.2
XM_017026412.1 159 UTR 5 XP_016881901.1

View Full Product Details