Product Details

SNP ID
rs146885610
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:42334150 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGTGCCTGCGAGCCGGGCTGGGG[C/G]GGTCCTGACTGTGGCCTGCAGGAGT
Phenotype
MIM: 604267
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
MEGF8 PubMed Links

Gene Details

Gene
MEGF8
Gene Name
multiple EGF like domains 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271938.1 1130 Silent Mutation GGC,GGG G165G NP_001258867.1
NM_001410.2 1130 Silent Mutation GGC,GGG G165G NP_001401.2
Gene
TMEM145
Gene Name
transmembrane protein 145
There are no transcripts associated with this gene.

View Full Product Details