Product Details
- SNP ID
-
rs146890196
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:42091302 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTCGGGCCCCCCTGGACCTCCAGG[A/G]GTCTGTGCTGCCGTCTCGCTGCAAG
- Phenotype
-
MIM: 164176
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
POU2F2
PubMed Links
Gene Details
- Gene
- POU2F2
- Gene Name
- POU class 2 homeobox 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001207025.2 |
2159 |
UTR 3 |
|
|
NP_001193954.1 |
NM_001207026.1 |
2159 |
Intron |
|
|
NP_001193955.1 |
NM_001247994.1 |
2159 |
UTR 3 |
|
|
NP_001234923.1 |
NM_002698.4 |
2159 |
UTR 3 |
|
|
NP_002689.1 |
XM_005259010.3 |
2159 |
Silent Mutation |
ACC,ACT |
T572T |
XP_005259067.1 |
XM_011527040.2 |
2159 |
Silent Mutation |
ACC,ACT |
T610T |
XP_011525342.1 |
XM_011527041.2 |
2159 |
UTR 3 |
|
|
XP_011525343.1 |
XM_011527042.2 |
2159 |
UTR 3 |
|
|
XP_011525344.1 |
XM_011527043.2 |
2159 |
UTR 3 |
|
|
XP_011525345.2 |
XM_017026884.1 |
2159 |
Silent Mutation |
ACC,ACT |
T709T |
XP_016882373.1 |
XM_017026885.1 |
2159 |
Silent Mutation |
ACC,ACT |
T709T |
XP_016882374.1 |
XM_017026886.1 |
2159 |
Silent Mutation |
ACC,ACT |
T697T |
XP_016882375.1 |
XM_017026887.1 |
2159 |
Silent Mutation |
ACC,ACT |
T697T |
XP_016882376.1 |
XM_017026888.1 |
2159 |
Silent Mutation |
ACC,ACT |
T697T |
XP_016882377.1 |
XM_017026889.1 |
2159 |
Silent Mutation |
ACC,ACT |
T697T |
XP_016882378.1 |
XM_017026890.1 |
2159 |
Silent Mutation |
ACC,ACT |
T697T |
XP_016882379.1 |
XM_017026891.1 |
2159 |
Silent Mutation |
ACC,ACT |
T693T |
XP_016882380.1 |
XM_017026892.1 |
2159 |
Silent Mutation |
ACC,ACT |
T687T |
XP_016882381.1 |
XM_017026893.1 |
2159 |
Silent Mutation |
ACC,ACT |
T594T |
XP_016882382.1 |
XM_017026894.1 |
2159 |
Silent Mutation |
ACC,ACT |
T588T |
XP_016882383.1 |
XM_017026895.1 |
2159 |
Silent Mutation |
ACC,ACT |
T588T |
XP_016882384.1 |
XM_017026896.1 |
2159 |
UTR 3 |
|
|
XP_016882385.1 |
- Gene
- ZNF574
- Gene Name
- zinc finger protein 574
There are no transcripts associated with this gene.
View Full Product Details