Product Details

SNP ID
rs146925848
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:8421915 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGGCAGCCCTGCCTTCTCCAAGGT[C/G]GTGGAGGCTACGGGCCTCGGACCGC
Phenotype
MIM: 613332
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MARCH2 PubMed Links

Gene Details

Gene
MARCH2
Gene Name
membrane associated ring-CH-type finger 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005415.1 245 Silent Mutation GTC,GTG V25V NP_001005415.1
NM_001005416.1 245 Silent Mutation GTC,GTG V25V NP_001005416.1
NM_016496.4 245 Silent Mutation GTC,GTG V25V NP_057580.3
XM_006722763.3 245 Silent Mutation GTC,GTG V25V XP_006722826.1
XM_011528046.2 245 Silent Mutation GTC,GTG V25V XP_011526348.1
XM_011528047.2 245 Silent Mutation GTC,GTG V25V XP_011526349.1
XM_017026853.1 245 Silent Mutation GTC,GTG V25V XP_016882342.1
XM_017026854.1 245 Intron XP_016882343.1

View Full Product Details