Product Details

SNP ID
rs147036683
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:54174182 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAGTGGATACAGAAGTAGATGGA[C/G]GCCCAGTACCGAAGGGTGTCGGCCA
Phenotype
MIM: 606048
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MBOAT7 PubMed Links

Gene Details

Gene
MBOAT7
Gene Name
membrane bound O-acyltransferase domain containing 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146056.2 1623 Silent Mutation GCC,GCG A354A NP_001139528.1
NM_001146082.2 1623 Intron NP_001139554.1
NM_001146083.2 1623 Silent Mutation GCC,GCG A354A NP_001139555.1
NM_024298.4 1623 Silent Mutation GCC,GCG A427A NP_077274.3
XM_011527299.2 1623 Silent Mutation GCC,GCG A427A XP_011525601.1
XM_011527300.2 1623 Silent Mutation GCC,GCG A427A XP_011525602.1
XM_017027296.1 1623 Intron XP_016882785.1
Gene
TMC4
Gene Name
transmembrane channel like 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145303.2 1623 Intron NP_001138775.2
NM_144686.3 1623 Intron NP_653287.2
XM_011526486.2 1623 Intron XP_011524788.1

View Full Product Details